3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR2097-1G OR2097-25G OR2097-5G
77406-588EA 48.9 USD
77406-588 77406-590 77406-592
3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid 98%
3-(2,4-Dimethylphenylcarbamoyl)phenylboronic acid
Formula: C₁₅H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD09027261
CAS Number: 957060-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR