4-Methyl-1H-pyrazole 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Fomepizole

OR480607-100G OR480607-25G
77410-018EA 206.82 USD
77410-018 77410-020
4-Methyl-1H-pyrazole 98%
4-Methyl-1H-pyrazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 99…100 °C/6 mmHg
Density: 0.99 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005245
CAS Number: 7554-65-6
UN: 1760
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR