3-Isopropylphenylboronic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Isopropylbenzeneboronic acid

OR10523-10G OR10523-5G
77402-760EA 186.44 USD
77402-760 77402-762
3-Isopropylphenylboronic acid 98%
3-Isopropylphenylboronic acid
Formula: C₉H₁₃BO₂
MW: 164.01 g/mol
Melting Pt: 72…76 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074634
CAS Number: 216019-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR