3-Nitrophenylsulfur pentafluoride 96+%

Supplier: APOLLO SCIENTIFIC
Warning

PC0472-1G PC0472-25G PC0472-5G
77412-574EA 315.96 USD
77412-574 77412-576 77412-578
3-Nitrophenylsulfur pentafluoride 96+%
3-Nitrophenylsulphur pentafluoride
Formula: C₆H₄F₅NO₂S
Boiling Pt: 106 °C/2 mmHg
Density: 1.662 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00221621
CAS Number: 2613-26-5
EINECS: 447-600-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR