Methyl-3-amino-5-hydroxybenzoate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 3-amino-5-hydroxybenzoate
OR12411-1G
OR12411-25G
OR12411-5G
77404-294EA
166.06
USD
77404-294
77404-296
77404-298
Methyl-3-amino-5-hydroxybenzoate
Methyl 3-amino-5-hydroxybenzoate
Formula:
C₈H₉NO₃ Melting Pt: 124…126 °C Storage Temperature: Ambient |
MDL Number:
MFCD09263220 CAS Number: 67973-80-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...