3-Chloro-5-iodobenzonitrile 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR15035-1G OR15035-25G OR15035-5G
77405-252EA 21.36 USD
77405-252 77405-254 77405-256
3-Chloro-5-iodobenzonitrile 99%
3-Chloro-5-iodobenzonitrile
Formula: C₇H₃ClIN
MW: 263.4629 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00672974
CAS Number: 289039-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR