2-Chloro-6-fluoro-3-nitrobenzonitrile

Supplier: APOLLO SCIENTIFIC
Danger

PC31540-1G PC31540-5G
77414-526EA 117.16 USD
77414-526 77414-528
2-Chloro-6-fluoro-3-nitrobenzonitrile
2-Chloro-6-fluoro-3-nitrobenzonitrile
Formula: C₇H₂ClFN₂O₂
Melting Pt: 91…93 °C
Storage Temperature: Ambient
MDL Number: MFCD00053834
CAS Number: 226419-18-7
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR