4-Bromo-3-methoxybenzamide 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR11558-1G
OR11558-250MG
77403-954EA
285.26
USD
77403-954
77403-956
4-Bromo-3-methoxybenzamide 98%
4-Bromo-3-methoxybenzamide
Formula:
C₈H₈BrNO₂ Storage Temperature: Ambient |
MDL Number:
MFCD09475887 CAS Number: 176961-57-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...