4'-Iodoacetanilide

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(4-Iodophenyl)acetamide

OR2193-1G OR2193-5G
77406-744EA 21.26 USD
77406-744 77406-746
4'-Iodoacetanilide
4'-Iodoacetanilide
Formula: C₈H₈INO
MW: 261.0597 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00016352
CAS Number: 622-50-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR