(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR22930-100G OR22930-1KG OR22930-500G
77406-974EA 105.96 USD
77406-974 77406-976 77406-978
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one 97+%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
Boiling Pt: 102…106 °C/0.25 mmHg
Melting Pt: 54…58 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00213364
CAS Number: 49805-30-3
EINECS: 421-830-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR