1-Methyl-1,4-cyclohexadiene, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

OR451306-100G OR451306-25G OR451306-5G
77409-784EA 352.52 USD
77409-784 77409-786 77409-788
1-Methyl-1,4-cyclohexadiene, Technical Grade
1-Methyl-1,4-cyclohexadiene
Formula: C₇H₁₀
MW: 94.16 g/mol
Boiling Pt: 114…115 °C
Density: 0.838 g/cm³ (25 °C)
Flash Pt: 11 °C
Storage Temperature: Ambient
MDL Number: MFCD00001538
CAS Number: 4313-57-9
UN: 3295
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR