2-Methoxy-5-(trifluoromethyl)benzonitrile 98%

Supplier: APOLLO SCIENTIFIC

PC302031-5G PC302031-1G PC302031-25G
77414-114EA 28.28 USD
77414-114 77414-110 77414-112
2-Methoxy-5-(trifluoromethyl)benzonitrile 98%
2-Methoxy-5-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
Melting Pt: 30…34 °C
Storage Temperature: Ambient
MDL Number: MFCD06409082
CAS Number: 34636-92-5
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR