tert-Butyldimethylsilyl trifluoromethanesulfonate 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
t-Butyldimethylsilyl trifluoromethanesulphonate
PC1605-5G
PC1605-100G
PC1605-25G
77413-274EA
21.32
USD
77413-274
77413-270
77413-272
tert-Butyldimethylsilyl trifluoromethanesulfonate 98%
tert-Butyldimethylsilyl trifluoromethanesulphonate
Electrophilic reagent for introduction of t-butyldimethylsilyl group. For a review see: Emde et al., Synthesis, 1982 , 1.
Formula:
C₇H₁₅F₃O₃SSi MW: 264.34 g/mol Boiling Pt: 65…67 °C/12 mmHg Density: 1.151 g/cm³ (25 °C) Flash Pt: 36 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000405 CAS Number: 69739-34-0 EINECS: 274-102-0 UN: 2920 ADR: 8+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...