Bis(tri-tert-butylphosphine)palladium(0)
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR17884-1G
OR17884-250MG
OR17884-5G
77406-144EA
58.08
USD
77406-144
77406-146
77406-148
Bis(tri-tert-butylphosphine)palladium(0)
Bis(tri-tert-butylphosphine)palladium(0)
Formula:
C₂₄H₅₄P₂Pd Melting Pt: 258…272 °C Storage Temperature: Freezer |
MDL Number:
MFCD03094580 CAS Number: 53199-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...