3-Cyano-4-fluorobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Fluoro-5-formylbenzonitrile

PC8847-100G PC8847-25G PC8847-5G
77418-468EA 114.1 USD
77418-468 77418-470 77418-472
3-Cyano-4-fluorobenzaldehyde 97%
3-Cyano-4-fluorobenzaldehyde
Formula: C₈H₄FNO
MW: 149.1219 g/mol
Melting Pt: 80…84 °C
Storage Temperature: Ambient
MDL Number: MFCD01863558
CAS Number: 218301-22-5
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR