2,6-Dichloro-5-fluoronicotinonitrile 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC9532-100G PC9532-25G
77418-676EA 50.94 USD
77418-676 77418-678
2,6-Dichloro-5-fluoronicotinonitrile 97%
2,6-Dichloro-5-fluoronicotinonitrile
Formula: C₆HCl₂FN₂
Melting Pt: 85…93 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01863289
CAS Number: 82671-02-1
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR