Benzofuran-5-carboxylic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR23098-1G OR23098-250MG
77407-100EA 37.2 USD
77407-100 77407-102
Benzofuran-5-carboxylic acid 95%
Benzofuran-5-carboxylic acid
Formula: C₉H₆O₃
MW: 162.1400 g/mol
Melting Pt: 189…190 °C
Storage Temperature: Ambient
MDL Number: MFCD01006742
CAS Number: 90721-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR