4-Amino-2-mercaptopyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine

OR1193T-1G OR1193T-250MG
77404-072EA 83.54 USD
77404-072 77404-074
4-Amino-2-mercaptopyrimidine
4-Amino-2-mercaptopyrimidine
Formula: C₄H₅N₃S
MW: 127.1676 g/mol
Melting Pt: 273…277 °C
Storage Temperature: Ambient
MDL Number: MFCD00006075
CAS Number: 333-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR