2,4,6-Trimethylheptane

Supplier: APOLLO SCIENTIFIC
Warning

OR2379-1G OR2379-250MG
77407-376EA 454.4 USD
77407-376 77407-378
2,4,6-Trimethylheptane
2,4,6-Trimethylheptane
Formula: C₁₀H₂₂
Boiling Pt: 144…147 °C
Density: 0.722 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 2613-61-8
UN: 3295
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR