4-Amino-3-methylbenzotrifluoride 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-4-(trifluoromethyl)aniline

PC9254-1G PC9254-250MG PC9254-5G
77418-630EA 41.78 USD
77418-630 77418-632 77418-634
4-Amino-3-methylbenzotrifluoride 97%
4-Amino-3-methylbenzotrifluoride
Formula: C₈H₈F₃N
MW: 175.15 g/mol
Boiling Pt: 98…99 °C/13 mmHg
Melting Pt: 20…25 °C
Density: 1.2 g/cm³ (25 °C)
Flash Pt: 84 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03701068
CAS Number: 67169-22-6
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR