N-Methyldicyclohexylamine 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR42279-100G OR42279-500G
77409-656EA 21.32 USD
77409-656 77409-658
N-Methyldicyclohexylamine 97%
N-Methyldicyclohexylamine
Formula: C₁₃H₂₅N
Boiling Pt: 265 °C
Density: 0.912 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00014289
CAS Number: 7560-83-0
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR