Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Ethyl 3-amino-4,4,4-trifluorobut-2-enoate
PC3161-100G
PC3161-25G
PC3161-500G
77414-536EA
52.98
USD
77414-536
77414-538
77414-540
Ethyl-3-amino-4,4,4-trifluorocrotonate 97%
Ethyl 3-amino-4,4,4-trifluorocrotonate
Formula:
C₆H₈F₃NO₂ MW: 183.13 g/mol Boiling Pt: 83…85 °C/15 mmHg Melting Pt: 24…26 °C Density: 1.245 g/cm³ (25 °C) Flash Pt: 65 °C Storage Temperature: Ambient |
MDL Number:
MFCD00068195 CAS Number: 372-29-2 UN: 3259 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...