3-Bromo-4-methylbenzaldehyde 98%

Supplier: APOLLO SCIENTIFIC

OR909388-100G OR909388-10G OR909388-25G
77411-584EA 425.86 USD
77411-584 77411-586 77411-588
3-Bromo-4-methylbenzaldehyde 98%
3-Bromo-4-methylbenzaldehyde
Formula: C₈H₇BrO
Storage Temperature: Ambient
MDL Number: MFCD04971197
CAS Number: 36276-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR