O-Isobutylhydroxylamine hydrochloride 98%

Supplier: APOLLO SCIENTIFIC

OR964831-5G OR964831-1G
77412-260EA 185.42 USD
77412-260 77412-258
O-Isobutylhydroxylamine hydrochloride 98%
O-Isobutylhydroxylamine hydrochloride
Formula: C₄H₁₂ClNO
MW: 125.6000 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00060208
CAS Number: 6084-58-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR