4-(Trifluoromethyl)cyclohexanone 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC7668N-100MG PC7668N-1G PC7668N-5G
77418-102EA 21.4 USD
77418-102 77418-104 77418-106
4-(Trifluoromethyl)cyclohexanone 97%
4-(Trifluoromethyl)cyclohexanone
Formula: C₇H₉F₃O
MW: 166.14
Boiling Pt: 67…68 °C/16 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00102145
CAS Number: 75091-99-5
UN: 1224
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR