4-Bromo-3-hydroxypyridine 95%

Supplier: APOLLO SCIENTIFIC
Warning

OR301092-10G OR301092-1G OR301092-5G
77408-768EA 308.7 USD
77408-768 77408-770 77408-772
4-Bromo-3-hydroxypyridine 95%
4-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MW: 173.9954 g/mol
Melting Pt: 134…136 °C
Storage Temperature: Ambient
MDL Number: MFCD03093021
CAS Number: 161417-28-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR