2,5-Dibromopyrazine 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR12499-100G OR12499-25G OR12499-5G
77404-326EA 379 USD
77404-326 77404-328 77404-330
2,5-Dibromopyrazine 97+%
2,5-Dibromopyrazine
Formula: C₄H₂Br₂N₂
Boiling Pt: 234 °C
Density: 2.197 g/cm³ (25 °C)
Flash Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD08061601
CAS Number: 23229-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR