2,4-Difluororesorcinol 97%

Supplier: APOLLO SCIENTIFIC

PC520578-1G PC520578-500MG
77416-506EA 192.56 USD
77416-506 77416-508
2,4-Difluororesorcinol 97%
2,4-Difluororesorcinol
Formula: C₆H₄F₂O₂
Storage Temperature: Freezer
CAS Number: 195136-71-1
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR