2,6-Diethylphenyl isocyanate 94%

Supplier: APOLLO SCIENTIFIC
Danger

OR33665-1G OR33665-25G OR33665-5G
77409-216EA 21.4 USD
77409-216 77409-218 77409-220
2,6-Diethylphenyl isocyanate 94%
2,6-Diethylphenyl isocyanate

  • May form small amount of sediment.

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00019912
CAS Number: 20458-99-5
UN: 2206
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR