Benzo[b]thiophene-7-carboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR12294-1G OR12294-250MG
77404-248EA 269.98 USD
77404-248 77404-250
Benzo[b]thiophene-7-carboxylic acid 97%
Benzo[b]thiophene-7-carboxylic acid
Formula: C₉H₆O₂S
Melting Pt: 176.5…178.5 °C
Storage Temperature: Refrigerator
CAS Number: 10134-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR