Benzo[b]thiophene-3-carboxylic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR23171-5G
OR23171-1G
OR23171-250MG
77407-154EA
186.44
USD
77407-154
77407-150
77407-152
Benzo[b]thiophene-3-carboxylic acid
Formula:
C₉H₆O₂S Melting Pt: 174…176 °C Storage Temperature: Ambient |
MDL Number:
MFCD01846406 CAS Number: 5381-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...