Acetoxyacetyl chloride
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR1899-100G
OR1899-25G
OR1899-5G
77406-428EA
126.34
USD
77406-428
77406-430
77406-432
Acetoxyacetyl chloride
Acetoxyacetyl chloride
Formula:
C₄H₅ClO₃ Boiling Pt: 55 °C/12 mmHg Density: 1.27 g/cm³ (25 °C) Flash Pt: 71 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011535 CAS Number: 13831-31-7 EINECS: 237-542-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...