1-(Trifluoroacetyl)imidazole 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC7240-25G PC7240-5G
77417-690EA 96.78 USD
77417-690 77417-692
1-(Trifluoroacetyl)imidazole 95%
1-(Trifluoroacetyl)imidazole

Derivatising agent. Useful for trifluoroacetylation ofalcohols and amines for electron-capture GC analysis.

Formula: C₅H₃F₃N₂O
Boiling Pt: 137…138 °C
Density: 1.441 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00014501
CAS Number: 1546-79-8
EINECS: 216-282-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR