Ethyl-4,4-difluoroacetoacetate 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl 4,4-difluoro-3-oxobutanoate, Ethyl 4,4-difluoro-3-oxobutyrate

PC5945-100G PC5945-25G PC5945-500G
77417-204EA 72.34 USD
77417-204 77417-206 77417-208
Ethyl-4,4-difluoroacetoacetate 96%
Ethyl-4,4-difluoroacetoacetate
Formula: C₆H₈F₂O₃
Boiling Pt: 162 °C
Density: 1.2418 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039277
CAS Number: 352-24-9
EINECS: 206-519-0
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR