Methyl-4,4,4-trifluorobutyrate

Supplier: APOLLO SCIENTIFIC
Warning

PC5516-1G PC5516-5G
77416-770EA 55.02 USD
77416-770 77416-772
Methyl-4,4,4-trifluorobutyrate
Methyl-4,4,4-trifluorobutyrate
Formula: C₅H₇F₃O₂
Boiling Pt: 113 °C
Storage Temperature: Ambient
CAS Number: 2365-82-4
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR