4-Chloro-3-(trifluoromethoxy)benzyl bromide 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Bromomethyl)-1-chloro-2-(trifluoromethoxy)benzene

PC49329-1G PC49329-25G PC49329-5G
77415-910EA 59.1 USD
77415-910 77415-912 77415-914
4-Chloro-3-(trifluoromethoxy)benzyl bromide 97%
4-Chloro-3-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD06660249
CAS Number: 886500-93-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR