4-(Trifluoromethoxy)toluene 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
PC7439W-100G
PC7439W-25G
PC7439W-5G
77417-972EA
191.14
USD
77417-972
77417-974
77417-976
4-(Trifluoromethoxy)toluene 97%
4-(Trifluoromethoxy)toluene
Formula:
C₈H₇F₃O Boiling Pt: 134…135 °C Density: 1.179 g/cm³ (25 °C) Flash Pt: 31 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042413 CAS Number: 706-27-4 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...