1-Chloro-2,6-difluorobenzene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC4958-25G PC4958-5G
77415-930EA 34.64 USD
77415-930 77415-932
1-Chloro-2,6-difluorobenzene 98%
1-Chloro-2,6-difluorobenzene
Formula: C₆H₃ClF₂
Boiling Pt: 131 °C
Density: 1.37 g/cm³ (25 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00142604
CAS Number: 38361-37-4
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR