3-Bromo-5-(trifluoromethyl)pyridine 98%

Supplier: APOLLO SCIENTIFIC
Danger

PC1078-1G PC1078-25G PC1078-5G
77412-980EA 21.26 USD
77412-980 77412-982 77412-984
3-Bromo-5-(trifluoromethyl)pyridine 98%
3-Bromo-5-(trifluoromethyl)pyridine
Formula: C₆H₃BrF₃N
Boiling Pt: 65…66 °C/19 mmHg
Melting Pt: 44…46 °C
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD04972700
CAS Number: 436799-33-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR