2'-Deoxyuridine
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0326-100G
OR0326-25G
OR0326-5G
77402-026EA
205.8
USD
77402-026
77402-028
77402-030
2'-Deoxyuridine
2'-Deoxyuridine
Formula:
C₉H₁₂N₂O₅ MW: 228.20 g/mol Melting Pt: 165…167 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006527 CAS Number: 951-78-0 EINECS: 213-455-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...