5-(Difluoromethoxy)-3-fluorobenzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC302611-1G PC302611-5G
77414-208EA 110.04 USD
77414-208 77414-210
5-(Difluoromethoxy)-3-fluorobenzyl bromide 95%
5-(Difluoromethoxy)-3-fluorobenzyl bromide
Formula: C₈H₆BrF₃O
MW: 255.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09832362
CAS Number: 1017779-39-3
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR