7,8-Dichloroisoquinoline 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR5292-1G OR5292-250MG
77410-268EA 132.38 USD
77410-268 77410-270
7,8-Dichloroisoquinoline 98%
7,8-Dichloroisoquinoline
Formula: C₉H₅Cl₂N
Melting Pt: 89…92 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258638
CAS Number: 61563-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR