4-(2-Chloroethyl)morpholine hydrochloride 99%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: N-(2-Chloroethyl)morpholine hydrochloride, N-(2-Chloroethyl)-Morpholinium Chloride

OR0990-100G OR0990-25G
77402-378EA 28.46 USD
77402-378 77402-380
4-(2-Chloroethyl)morpholine hydrochloride 99%
4-(2-Chloroethyl)morpholine hydrochloride
Formula: C₆H₁₃Cl₂NO
MW: 186.08 g/mol
Melting Pt: 180…186 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012797
CAS Number: 3647-69-6
EINECS: 222-881-2
UN: 2923
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR