2-Bromo-5-methoxyphenylboronic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-5-methoxybenzeneboronic acid, 2-Bromo-5-methoxybenzene boronic acid

OR1188-1G OR1188-250MG
77404-048EA 90.68 USD
77404-048 77404-050
2-Bromo-5-methoxyphenylboronic acid
2-Bromo-5-methoxyphenylboronic acid
Formula: C₇H₈BBrO₃
MW: 230.85 g/mol
Melting Pt: 214…216 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06659859
CAS Number: 89694-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR