2-Chloro-4-fluorophenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-(2-Chloro-4-fluorophenyl)acetonitrile

PC4093-1G PC4093-5G
77415-124EA 21.26 USD
77415-124 77415-126 77415-128
2-Chloro-4-fluorophenylacetonitrile 97%
2-Chloro-4-fluorophenylacetonitrile
Formula: C₈H₅ClFN
MW: 169.59 g/mol
Melting Pt: 67…69 °C
Storage Temperature: Ambient
MDL Number: MFCD00236029
CAS Number: 75279-56-0
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR