Bis(2-chloroethyl)ammonium chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Bis(2-chloroethyl)amine hydrochloride

OR27981-1KG OR27981-250G OR27981-10KG
77408-014EA 66.22 USD
77408-014 77408-016 77408-012
Bis(2-chloroethyl)ammonium chloride 98%
Bis(2-chloroethyl)ammonium chloride
Formula: C₄H₁₀Cl₃N
MW: 178.49 g/mol
Melting Pt: 211…215 °C
Storage Temperature: Ambient
MDL Number: MFCD00012515
CAS Number: 821-48-7
EINECS: 212-479-5
UN: 2923
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR