Potassium methyl malonate 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt

OR25586-100G OR25586-250G OR25586-500G
77407-614EA 33.62 USD
77407-614 77407-616 77407-618
Potassium methyl malonate 98%
Potassium methyl malonate
Formula: C₄H₅KO₄
MW: 156.18
Melting Pt: 198…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00014021
CAS Number: 38330-80-2
EINECS: 253-886-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR