4-Nitrobiphenyl 95%

Supplier: APOLLO SCIENTIFIC
Danger

OR26189-1G OR26189-5G
77407-748EA 32.6 USD
77407-748 77407-750
4-Nitrobiphenyl 95%
4-Nitrobiphenyl
Formula: C₁₂H₉NO₂
Boiling Pt: 340 °C
Melting Pt: 111…115 °C
Storage Temperature: Ambient
CAS Number: 92-93-3
EINECS: 202-204-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR