Thiophene-2-carbonyl chloride 98%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Thiophenecarbonyl chloride, Thiophene-2-carboxylic acid chloride

OR23195-100G OR23195-500G
77407-168EA 41.78 USD
77407-168 77407-170
Thiophene-2-carbonyl chloride 98%
Thiophene-2-carbonyl chloride
Formula: C₅H₃ClOS
MW: 146.6 g/mol
Boiling Pt: 206…208 °C
Density: 1.371 g/cm³ (25 °C)
Flash Pt: 91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005428
CAS Number: 5271-67-0
EINECS: 226-092-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR