1H-Pyrazole-5-carbaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR301050-100G OR301050-25G OR301050-5G
77408-740EA 509.4 USD
77408-740 77408-742 77408-744
1H-Pyrazole-5-carbaldehyde 97%
Formula: C₄H₄N₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06657574
CAS Number: 948552-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR